Learn More
V&P Scientific Inc MICROPLATE, 96 Deep Well Format, 2mL Round Well, SLAS Footprint, PTFE
Catalog No. NC2233222
Encompass
MICROPLATE, 96 Deep Well Format, 8mm Diameter, 2mL Volume, Round Well, SLAS Footprint, PTFE
This item is not returnable. View return policy
Catalog No. NC2233222 Supplier V&P Scientific Inc Supplier No. VP 416DPTFE
MICROPLATE, 96 Deep Well Format, 8mm Diameter, 2mL Volume, Round Well, SLAS Footprint, PTFE