Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, 0.5 mL per Well (V-conical, sterile)
SDP
Catalog No. 502733180
Encompass_Preferred
The Chemglass CLS-3506-S17 is a sterile, deep 96-well microplate with 0.5 mL capacity per well and V-conical bottoms, designed for high-throughput workflows, sample storage, and nucleic acid extraction. It is compatible with multi-channel pipettes and automated equipment.
  • 0.5 mL capacity per well.
  • V-conical bottom for efficient sample recovery.
  • Deep-well, 96-well format suitable for storage and processing.
  • Sterile, single-use format.
  • Polypropylene construction for chemical resistance (inferred from product family).
  • Compatible with multi-channel pipettes and automated equipment.
Catalog No. 50-273-3180 Supplier Chemglass Life Sciences Supplier No. CLS3506S17
Add to Cart
Add to Cart
The Chemglass CLS-3506-S17 is a sterile, deep 96-well microplate with 0.5 mL capacity per well and V-conical bottoms, designed for high-throughput workflows, sample storage, and nucleic acid extraction. It is compatible with multi-channel pipettes and automated equipment.
  • 0.5 mL capacity per well.
  • V-conical bottom for efficient sample recovery.
  • Deep-well, 96-well format suitable for storage and processing.
  • Sterile, single-use format.
  • Polypropylene construction for chemical resistance (inferred from product family).
  • Compatible with multi-channel pipettes and automated equipment.