Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well, 0.5 mL deep well, V-conical bottom, non-sterile, square well top (CLS-3506-15)
SDP
Catalog No. 502733191
Encompass_Preferred
This 96-well deep microplate provides 0.5 mL well capacity with V-conical bottoms and square well tops, designed for high-throughput workflows such as nucleic acid extraction and sample storage. The plate meets SBS/ANSI footprint dimensions and is compatible with multichannel pipettes and automated equipment.
  • 96-well deep well format with 0.5 mL capacity per well.
  • V conical bottom to minimize residual volume.
  • Square well top compatible with multichannel pipettes and automation.
  • Temperature range -80°C to 120°C; autoclavable.
  • Centrifuge rated up to 3,000 g.
  • SBS/ANSI footprint and stackable design for automated workflows.
  • Alphanumeric well coding for easy identification.
  • Non-pyrogenic, DNase/RNase free; non-sterile.
  • Sold in cases of five (CS5).
Catalog No. 50-273-3191 Supplier Chemglass Life Sciences Supplier No. CLS350615
Add to Cart
Add to Cart
This 96-well deep microplate provides 0.5 mL well capacity with V-conical bottoms and square well tops, designed for high-throughput workflows such as nucleic acid extraction and sample storage. The plate meets SBS/ANSI footprint dimensions and is compatible with multichannel pipettes and automated equipment.
  • 96-well deep well format with 0.5 mL capacity per well.
  • V conical bottom to minimize residual volume.
  • Square well top compatible with multichannel pipettes and automation.
  • Temperature range -80°C to 120°C; autoclavable.
  • Centrifuge rated up to 3,000 g.
  • SBS/ANSI footprint and stackable design for automated workflows.
  • Alphanumeric well coding for easy identification.
  • Non-pyrogenic, DNase/RNase free; non-sterile.
  • Sold in cases of five (CS5).