Learn More
Chemglass Life Sciences Microplate, 96-Well, 0.5mL, Deep, Non-Sterile, Round Well Top
SDP
Catalog No. 5000980741
Encompass_Preferred
Our deep well micro plates are manufactured from USP grade class VI polypropylene material and feature uniform external dimensions and tolerances to meet SBS recommendations for compatibility with equipment and robotics. Micro plates are biologically inert. The material properties provide optimal conditions for laboratory specimen and compound storage. Ideal for cultivating bacteria or yeasts. Micro plates feature alphanumerically coded wells. Conical well bottom design aids in complete sample recovery. Plates are autoclavable and supplied without lids.
Catalog No. 50-009-80741 Supplier Chemglass Life Sciences Supplier No. CLS350601
Our deep well micro plates are manufactured from USP grade class VI polypropylene material and feature uniform external dimensions and tolerances to meet SBS recommendations for compatibility with equipment and robotics. Micro plates are biologically inert. The material properties provide optimal conditions for laboratory specimen and compound storage. Ideal for cultivating bacteria or yeasts. Micro plates feature alphanumerically coded wells. Conical well bottom design aids in complete sample recovery. Plates are autoclavable and supplied without lids.