Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, 1.0 mL, Sterile, Round Well (CLS-3506-S3)
SDP
Catalog No. 502733175
Encompass_Preferred
Deep-well 96-well microplate designed for sample storage, culture, and automated handling. Manufactured from USP Class VI polypropylene with uniform external dimensions to meet SBS standards, the plate features alphanumeric well coding and conical well bottoms to aid sample recovery. Plates are autoclavable and supplied without lids.
  • 96 wells with 1.0 mL nominal well volume.
  • USP Class VI polypropylene construction.
  • Round well top with conical bottom for efficient sample recovery.
  • Alphanumeric well coding for well identification.
  • Autoclavable and compatible with automation and robotic systems.
  • Supplied sterile and without lids.
Catalog No. 50-273-3175 Supplier Chemglass Life Sciences Supplier No. CLS3506S3
Add to Cart
Add to Cart
Deep-well 96-well microplate designed for sample storage, culture, and automated handling. Manufactured from USP Class VI polypropylene with uniform external dimensions to meet SBS standards, the plate features alphanumeric well coding and conical well bottoms to aid sample recovery. Plates are autoclavable and supplied without lids.
  • 96 wells with 1.0 mL nominal well volume.
  • USP Class VI polypropylene construction.
  • Round well top with conical bottom for efficient sample recovery.
  • Alphanumeric well coding for well identification.
  • Autoclavable and compatible with automation and robotic systems.
  • Supplied sterile and without lids.