Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well, 1.0 mL, Deep, Round Well Top
SDP
Catalog No. 502733195
Encompass_Preferred
Chemglass 96-well deep microplate with 1.0 mL per well, designed for sample storage, cultivation, and automated workflows. Constructed from USP Class VI polypropylene to meet SBS/ANSI dimensions, the plate is autoclavable and compatible with multichannel pipettes and robotic systems.
  • 96 wells with 1.0 mL capacity per well.
  • Round well top with deep U-bottom design.
  • USP Class VI polypropylene material; biologically inert.
  • Autoclavable; temperature range -80°C to 120°C.
  • Centrifugal strength up to 3,000 g.
  • Stackable design with alphanumeric well coding for easy identification.
  • Supplied non-sterile; packaged as case of 5 (supplier listing).
Catalog No. 50-273-3195 Supplier Chemglass Life Sciences Supplier No. CLS350603
Add to Cart
Add to Cart
Chemglass 96-well deep microplate with 1.0 mL per well, designed for sample storage, cultivation, and automated workflows. Constructed from USP Class VI polypropylene to meet SBS/ANSI dimensions, the plate is autoclavable and compatible with multichannel pipettes and robotic systems.
  • 96 wells with 1.0 mL capacity per well.
  • Round well top with deep U-bottom design.
  • USP Class VI polypropylene material; biologically inert.
  • Autoclavable; temperature range -80°C to 120°C.
  • Centrifugal strength up to 3,000 g.
  • Stackable design with alphanumeric well coding for easy identification.
  • Supplied non-sterile; packaged as case of 5 (supplier listing).