Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep U-Bottom, 1.3 mL per Well, Sterile (CLS-3506-S8)
SDP
Catalog No. 502733167
Encompass_Preferred
This 96-well deep microplate provides 1.3 mL capacity per well in an SBS/ANSI-format plate designed for deep-well storage, sample processing, and automated workflows. Available sterile in case quantities, the plates are made from USP grade class VI polypropylene and are compatible with multichannel pipettes and common automation platforms.
  • 96 wells.
  • 1.3 mL well volume.
  • U-bottom, round well top.
  • USP grade class VI polypropylene material.
  • Temperature range -80°C to 120°C.
  • Sterile variant available; select variants autoclavable.
  • DNase/RNase free and non-pyrogenic.
  • Compatible with SBS/ANSI footprint and automated equipment.
  • Pack quantity: case of 5 for sterile packs.
Catalog No. 50-273-3167 Supplier Chemglass Life Sciences Supplier No. CLS3506S8
Add to Cart
Add to Cart
This 96-well deep microplate provides 1.3 mL capacity per well in an SBS/ANSI-format plate designed for deep-well storage, sample processing, and automated workflows. Available sterile in case quantities, the plates are made from USP grade class VI polypropylene and are compatible with multichannel pipettes and common automation platforms.
  • 96 wells.
  • 1.3 mL well volume.
  • U-bottom, round well top.
  • USP grade class VI polypropylene material.
  • Temperature range -80°C to 120°C.
  • Sterile variant available; select variants autoclavable.
  • DNase/RNase free and non-pyrogenic.
  • Compatible with SBS/ANSI footprint and automated equipment.
  • Pack quantity: case of 5 for sterile packs.