Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, U-Bottom (1.6 mL wells) - CLS-3506-S12
SDP
Catalog No. 502733183
Encompass_Preferred
The Chemglass CLS-3506-S12 is a sterile, 96-well deep microplate with U-bottom wells and a square well top, designed for sample storage and high-throughput workflows. Wells hold 1.6 mL and the plate is compatible with automated liquid handling and multi-channel pipettes.
  • 96 wells with 1.6 mL well volume.
  • U-bottom well geometry with square well top.
  • Sterile and DNase/RNase free.
  • Compatible with multi-channel pipettes and automated equipment.
  • Temperature range: -80°C to 120°C.
  • Autoclavable; centrifugal strength up to 3000 g.
  • Conforms to SBS/ANSI 96-well footprint for automation compatibility.
  • Stackable design with reliable tight sealing.
Catalog No. 50-273-3183 Supplier Chemglass Life Sciences Supplier No. CLS3506S12
Add to Cart
Add to Cart
The Chemglass CLS-3506-S12 is a sterile, 96-well deep microplate with U-bottom wells and a square well top, designed for sample storage and high-throughput workflows. Wells hold 1.6 mL and the plate is compatible with automated liquid handling and multi-channel pipettes.
  • 96 wells with 1.6 mL well volume.
  • U-bottom well geometry with square well top.
  • Sterile and DNase/RNase free.
  • Compatible with multi-channel pipettes and automated equipment.
  • Temperature range: -80°C to 120°C.
  • Autoclavable; centrifugal strength up to 3000 g.
  • Conforms to SBS/ANSI 96-well footprint for automation compatibility.
  • Stackable design with reliable tight sealing.