Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, 2.0 mL, Sterile, Square Well Top
SDP
Catalog No. 502733170
Encompass_Preferred
The Chemglass 96-well deep well microplate provides 2.0 mL well capacity for sample storage and processing. It is manufactured from USP grade Class VI polypropylene for laboratory compatibility and is designed to work with automated equipment and multi-channel pipettes.
  • Made from USP grade Class VI polypropylene.
  • Well volume: 2.0 mL per well.
  • Well top: Square well top.
  • Sterility: Supplied sterile.
  • Autoclavable: Yes.
  • Alphanumeric coding: Features alphanumerically coded wells for easy identification.
  • Dimensions: Uniform external dimensions and tolerances to meet SBS recommendations.
Catalog No. 50-273-3170 Supplier Chemglass Life Sciences Supplier No. CLS3506S5
Add to Cart
Add to Cart
The Chemglass 96-well deep well microplate provides 2.0 mL well capacity for sample storage and processing. It is manufactured from USP grade Class VI polypropylene for laboratory compatibility and is designed to work with automated equipment and multi-channel pipettes.
  • Made from USP grade Class VI polypropylene.
  • Well volume: 2.0 mL per well.
  • Well top: Square well top.
  • Sterility: Supplied sterile.
  • Autoclavable: Yes.
  • Alphanumeric coding: Features alphanumerically coded wells for easy identification.
  • Dimensions: Uniform external dimensions and tolerances to meet SBS recommendations.