Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, 2.0 mL per Well (CLS-3506-05)
SDP
Catalog No. 502733194
Encompass_Preferred
The Chemglass 96-well deep-well microplate is a polypropylene plate designed for sample storage, cultivation, and high-throughput workflows. It provides 2.0 mL capacity per well, SBS-compatible external dimensions, and a conical well bottom to aid complete sample recovery.
  • Manufactured from USP grade Class VI polypropylene.
  • 2.0 mL nominal volume per well.
  • Conical (pyramid) well bottom for efficient sample recovery.
  • Alphanumerically coded wells for sample identification.
  • Compatible with SBS-standard automation and equipment.
  • Autoclavable; supplied non-sterile.
  • Packaged as CS5 (case of 5).
Catalog No. 50-273-3194 Supplier Chemglass Life Sciences Supplier No. CLS350605
Add to Cart
Add to Cart
The Chemglass 96-well deep-well microplate is a polypropylene plate designed for sample storage, cultivation, and high-throughput workflows. It provides 2.0 mL capacity per well, SBS-compatible external dimensions, and a conical well bottom to aid complete sample recovery.
  • Manufactured from USP grade Class VI polypropylene.
  • 2.0 mL nominal volume per well.
  • Conical (pyramid) well bottom for efficient sample recovery.
  • Alphanumerically coded wells for sample identification.
  • Compatible with SBS-standard automation and equipment.
  • Autoclavable; supplied non-sterile.
  • Packaged as CS5 (case of 5).