Learn More
Chemglass Life Sciences Microplate, 96-Well Deep, 2.2 mL per Well, U-Bottom, Square Well Top, Non-Sterile (CS5)
SDP
Catalog No. 502733193
Encompass_Preferred
The Chemglass Microplate CLS-3506-13 is a 96-well deep microplate with 2.2 mL capacity per well, designed for storage, sample processing, and nucleic acid extraction workflows. It is non-sterile and supplied in casepack CS5. The plate conforms to SBS/ANSI footprints and is compatible with multichannel pipettes and automated equipment.
  • 2.2 mL capacity per well.
  • U-bottom wells with a square well top.
  • Non-sterile supply.
  • Pack quantity: CS5.
  • Temperature range: -80°C to 120°C.
  • Autoclavable.
  • Centrifugal strength up to 3,000 g.
  • Compatible with multichannel pipettes and automation equipment.
  • Conforms to SBS/ANSI 96-well footprint.
  • Optimized bottom design to minimize residual volume.
  • Clear alphanumeric well codes for identification.
  • Stackable with reliable sealing.
  • Non-pyrogenic and DNase/RNase free.
Catalog No. 50-273-3193 Supplier Chemglass Life Sciences Supplier No. CLS350613
Add to Cart
Add to Cart
The Chemglass Microplate CLS-3506-13 is a 96-well deep microplate with 2.2 mL capacity per well, designed for storage, sample processing, and nucleic acid extraction workflows. It is non-sterile and supplied in casepack CS5. The plate conforms to SBS/ANSI footprints and is compatible with multichannel pipettes and automated equipment.
  • 2.2 mL capacity per well.
  • U-bottom wells with a square well top.
  • Non-sterile supply.
  • Pack quantity: CS5.
  • Temperature range: -80°C to 120°C.
  • Autoclavable.
  • Centrifugal strength up to 3,000 g.
  • Compatible with multichannel pipettes and automation equipment.
  • Conforms to SBS/ANSI 96-well footprint.
  • Optimized bottom design to minimize residual volume.
  • Clear alphanumeric well codes for identification.
  • Stackable with reliable sealing.
  • Non-pyrogenic and DNase/RNase free.