Learn More
Chemglass Life Sciences Chemglass Microplate, 96-Well Deep, 2.2 mL per Well, U-Bottom, Sterile
SDP
Catalog No. 502733182
Encompass_Preferred
96-well deep microplate with 2.2 mL well volume, U-bottom wells and a square well top. Designed to SBS/ANSI 96-well format specifications for compatibility with multichannel pipettes and automated equipment. Sterile, stackable, and DNase/RNase free; supports -80°C to 120°C and is autoclavable.
  • 96 wells, 2.2 mL capacity per well.
  • U-bottom wells with square well top.
  • Sterile; DNase/RNase free; non-pyrogenic.
  • SBS/ANSI footprint for automation compatibility.
  • Temperature range -80°C to 120°C.
  • Autoclavable; centrifuge up to 3,000 x g.
  • Stackable with reliable tight sealing.
  • Optimized bottom design to minimize residual volume.
  • Clear alphanumeric well identification for easy tracking.
Catalog No. 50-273-3182 Supplier Chemglass Life Sciences Supplier No. CLS3506S13
Add to Cart
Add to Cart
96-well deep microplate with 2.2 mL well volume, U-bottom wells and a square well top. Designed to SBS/ANSI 96-well format specifications for compatibility with multichannel pipettes and automated equipment. Sterile, stackable, and DNase/RNase free; supports -80°C to 120°C and is autoclavable.
  • 96 wells, 2.2 mL capacity per well.
  • U-bottom wells with square well top.
  • Sterile; DNase/RNase free; non-pyrogenic.
  • SBS/ANSI footprint for automation compatibility.
  • Temperature range -80°C to 120°C.
  • Autoclavable; centrifuge up to 3,000 x g.
  • Stackable with reliable tight sealing.
  • Optimized bottom design to minimize residual volume.
  • Clear alphanumeric well identification for easy tracking.