Learn More
Chemglass Life Sciences Chemglass microplate, 96-well deep, 2.2 mL wells, U-bottom, sterile
SDP
Catalog No. 502733179
Encompass_Preferred
Deep-well 96-well microplate with 2.2 mL capacity per well, designed for sample storage and processing. The plate is sterile, autoclavable, and compatible with refrigeration to -80°C; it withstands moderate centrifugal forces for routine laboratory workflows.
  • 96 wells.
  • 2.2 mL well volume.
  • U-bottom with square well top.
  • Sterile.
  • Autoclavable.
  • Temperature range: -80°C to 120°C.
  • Centrifugation rating up to 3,000 g.
Catalog No. 50-273-3179 Supplier Chemglass Life Sciences Supplier No. CLS3506S19
Deep-well 96-well microplate with 2.2 mL capacity per well, designed for sample storage and processing. The plate is sterile, autoclavable, and compatible with refrigeration to -80°C; it withstands moderate centrifugal forces for routine laboratory workflows.
  • 96 wells.
  • 2.2 mL well volume.
  • U-bottom with square well top.
  • Sterile.
  • Autoclavable.
  • Temperature range: -80°C to 120°C.
  • Centrifugation rating up to 3,000 g.