Learn More
Chemglass Life Sciences Microplate, 96-Well Deep, 2.2 mL per Well, V Conical Bottom, Square Well Top, Non-Sterile
SDP
Catalog No. 502733192
Encompass_Preferred
This 96-well deep microplate provides 2.2 mL capacity per well with V-conical bottoms and square well tops, designed for sample storage, processing, and compatibility with automated lab equipment. It is non-sterile and conforms to SBS/ANSI 96-well dimensions for use with multi-channel pipettes and automation.
  • 2.2 mL capacity per well.
  • V-conical bottom and square well top to minimize residual volume.
  • Non-sterile.
  • Autoclavable; recommended temperature range -80°C to 120°C.
  • Centrifugal strength up to 3,000 g.
  • Compatible with multi-channel pipettes and automated equipment.
  • Optimized bottom design to reduce residual volume.
  • Clear alphanumeric codes for easy identification.
  • Stackable with reliable sealing.
  • Non-pyrogenic, DNase/RNase free.
Catalog No. 50-273-3192 Supplier Chemglass Life Sciences Supplier No. CLS350614
Add to Cart
Add to Cart
This 96-well deep microplate provides 2.2 mL capacity per well with V-conical bottoms and square well tops, designed for sample storage, processing, and compatibility with automated lab equipment. It is non-sterile and conforms to SBS/ANSI 96-well dimensions for use with multi-channel pipettes and automation.
  • 2.2 mL capacity per well.
  • V-conical bottom and square well top to minimize residual volume.
  • Non-sterile.
  • Autoclavable; recommended temperature range -80°C to 120°C.
  • Centrifugal strength up to 3,000 g.
  • Compatible with multi-channel pipettes and automated equipment.
  • Optimized bottom design to reduce residual volume.
  • Clear alphanumeric codes for easy identification.
  • Stackable with reliable sealing.
  • Non-pyrogenic, DNase/RNase free.