Learn More
Chemglass Life Sciences Microplate, 96-Well, 2.2mL, Deep Square, 44mm Height, Pyramid Bottom, Toughened for Genomics, Porvair
SDP
Catalog No. 5000980713
Encompass_Preferred
Made from virgin polypropylene. Tested for low extractables. V bottom to allow total liquid removal, partial collection and to aid re-suspension. No inner edges to allow better collection of magnetic beads. DNase/ RNase free. Working volumes of 2 and 2. 2mL. Raised well rims to improve heat sealing. Conical base which aids sample concentration, reconstitution and centrifugation. Toughened genomics version for seed and leaf grinding. 96 square well plates designed so that they would make the interchange of plates simpler in automated systems. Plates have the same geometry and the only variable is the well depth.
Catalog No. 50-009-80713 Supplier Chemglass Life Sciences Supplier No. CLS3560022
Made from virgin polypropylene. Tested for low extractables. V bottom to allow total liquid removal, partial collection and to aid re-suspension. No inner edges to allow better collection of magnetic beads. DNase/ RNase free. Working volumes of 2 and 2. 2mL. Raised well rims to improve heat sealing. Conical base which aids sample concentration, reconstitution and centrifugation. Toughened genomics version for seed and leaf grinding. 96 square well plates designed so that they would make the interchange of plates simpler in automated systems. Plates have the same geometry and the only variable is the well depth.