Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices Microplate, 384-Well, Large Volume, With Cover

Catalog No. 5000014355
Encompass_Preferred

The Molecular Devices 384-well large-volume microplate provides increased per-well capacity for high-throughput assays and is supplied with a fitted cover for standard plate handling and instrument compatibility.

  • Well count: 384.
  • Maximum well volume: 95 μL.
  • Well type: large volume.
  • External dimensions without lid: 127.9 x 86 x 10 mm.
  • External dimensions with lid: 128 x 86 x 12.5 mm.
  • Includes fitted cover.

Catalog No. 50-000-14355 Supplier Molecular Devices Supplier No. X7001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Molecular Devices 384-well large-volume microplate provides increased per-well capacity for high-throughput assays and is supplied with a fitted cover for standard plate handling and instrument compatibility.

  • Well count: 384.
  • Maximum well volume: 95 μL.
  • Well type: large volume.
  • External dimensions without lid: 127.9 x 86 x 10 mm.
  • External dimensions with lid: 128 x 86 x 12.5 mm.
  • Includes fitted cover.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.