Learn More
USA Scientific Inc Microplate film, sterile
Catalog No. NC9957101
Encompass
Microplate film, polyester-based, -40C to +120C, sterile, 100/box
Catalog No. NC9957101 Supplier USA Scientific Inc Supplier No. 29200010
Microplate film, polyester-based, -40C to +120C, sterile, 100/box