Learn More
BioSpec Products MICROPLATE BEAD LOADER 1PK
SDP
Catalog No. NC0501501
Encompass_Preferred
96-well manual microplate bead loader. Loads 1 ml of beads into all 96 wells of a 2 ml deep well microplate at one time. 1 ea
This item is not returnable. View return policy
Catalog No. NC0501501 Supplier BioSpec Products Supplier No. 702L
96-well manual microplate bead loader. Loads 1 ml of beads into all 96 wells of a 2 ml deep well microplate at one time. 1 ea