Learn More
Andwin Scientific MICROPLATE BLOCK
SDP
Catalog No. NC9461426
Encompass_Preferred
Modular Heating Block, Titer Plate. Solid aluminum, anodized blue modular block for producing maximum heat retention in and Signature Dry 2/4/6 Block Heaters. Equipped with thermometer well.
Catalog No. NC9461426 Supplier Andwin Scientific Supplier No. 13259295
Modular Heating Block, Titer Plate. Solid aluminum, anodized blue modular block for producing maximum heat retention in and Signature Dry 2/4/6 Block Heaters. Equipped with thermometer well.