Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates cellGrade™ 384-Well Flat-Bottom Microplates, PS

Catalog No. 14386046 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-386-046 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-386-046 Supplier BRANDTECH™ BRAND™ Supplier No. 781982
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile 384-well microplates with cellGrade™ surface treatment for optimal growth. Ideal for cultivating adherent cells with a hydrophilic PS surface featuring accessible carboxyl and hydroxyl groups. Enhances serum component binding for effective indirect cell adhesion.

  • Polystyrene construction
  • Sterile with included lids
  • Designed for the cultivation of adherent cell lines
  • Hydrophilic PS surface with accessible carboxyl and hydroxyl groups
  • Enhance serum component binding for indirect cell adhesion
  • Clearly distinguishable with orange color code and embossed alphanumeric coding for 384-well plates
  • BIO-CERT™ CELL CULTURE QUALITY: free from endotoxins, DNase, DNA, RNase, non-cytotoxic
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ANSI, SLAS
Lid With Lid
Array 16 x 24
Autoclavable Not Autoclavable
Binding Type Carboxyl and Hydroxyl Groups
Product Line BRANDplates
Color Black
Product Type 384-well microplate
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment cellGrade
Total Volume (Metric) Well 100 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Cultivation of adherent cell lines
No. per Case 50/Cs.
No. per Pack 50
RNase Free Yes
Fit for Purpose Sterile
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.