Learn More
BioSpec Products 2 ml Deep well Microplate, 50 plates per case
SDP
Catalog No. NC1428962
Encompass_Preferred
Porvair Science's reinforced polypropylene, 2 ml square 96 deep well microplates have extra structural ribbing on their bottom which provides support when used with a high energy shaking bead mill. Their microplates are made of specially formulated polypropylene and pair with special reusable mats. The microplates can also be sealed with adhesive-bonded or heat-bonded films, but these films seldom prevent leaking during intense shaking in a Mini-BeadBeater-96.
Catalog No. NC1428962 Supplier BioSpec Products Supplier No. 219030
Porvair Science's reinforced polypropylene, 2 ml square 96 deep well microplates have extra structural ribbing on their bottom which provides support when used with a high energy shaking bead mill. Their microplates are made of specially formulated polypropylene and pair with special reusable mats. The microplates can also be sealed with adhesive-bonded or heat-bonded films, but these films seldom prevent leaking during intense shaking in a Mini-BeadBeater-96.