Learn More
Waters Corp Waters Pipette, Single-channel, 0.2–10 μL, Bluetooth
Catalog No. 502269330 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Andrew Alliance Bluetooth Electronic Single-channel Pipettes provide accurate, precise, and ergonomic pipetting. They are compatible with Andrew+ and Pipette+ systems for guided or robotic use, and OneLab software enables remote programming and traceability. Electronic operation reduces user variability and features an ergonomic design to prevent repetitive strain injuries.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more
  • Volume range: 0.2–10 μL
  • Number of channels: 1
  • Volume increment: 0.01
  • Compatible with Andrew+ and Pipette+ systems
  • Compatible with OneLab software
This item is not returnable. View return policy
Catalog No. 50-226-9330 Supplier Waters Corp Supplier No. 186009769
Andrew Alliance Bluetooth Electronic Single-channel Pipettes provide accurate, precise, and ergonomic pipetting. They are compatible with Andrew+ and Pipette+ systems for guided or robotic use, and OneLab software enables remote programming and traceability. Electronic operation reduces user variability and features an ergonomic design to prevent repetitive strain injuries.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more
  • Volume range: 0.2–10 μL
  • Number of channels: 1
  • Volume increment: 0.01
  • Compatible with Andrew+ and Pipette+ systems
  • Compatible with OneLab software