Learn More
Alkali Scientific ASI™ Microplate Lids for Microplates, Polystyrene, Non-Sterile, 150/Cs
SDP
Catalog No. 503027439
Encompass_Preferred
96-well microtest plates for general (non-tissue culture) applications including microbiology and serology.Available in “U” (round bottom), “V” (conical bottom) and fl at bottom shapes. Each plate is individually wrapped to protect from air contaminants. Made from high optic transmission grade polystyrene, the microtest plates are arranged in a standard 12 x 8 configuration and have alphanumeric position markings. Optional lids are also available. Both plates and lids may be purchased sterile or non-sterile.
Catalog No. 50-302-7439 Supplier Alkali Scientific Supplier No. MPL25
96-well microtest plates for general (non-tissue culture) applications including microbiology and serology.Available in “U” (round bottom), “V” (conical bottom) and fl at bottom shapes. Each plate is individually wrapped to protect from air contaminants. Made from high optic transmission grade polystyrene, the microtest plates are arranged in a standard 12 x 8 configuration and have alphanumeric position markings. Optional lids are also available. Both plates and lids may be purchased sterile or non-sterile.