Learn More
LUNA NANOTECH INC Microplate Magnetic Separation Rack
Catalog No. 502566968
Encompass_Preferred
96-well Microplate Magnetic Separation Rack
- Designed to be used with 96-well microplates, ELISA plates
- 96 magnets are positiont below microplate wells for efficient magnetic separation
- Can be used for high throughput separation of up to 96 samples
- Housing material: PMMA/Acrylic
- Magnet: N38 Neodymium (NdFeB)
Catalog No. 50-256-6968 Supplier LUNA NANOTECH INC Supplier No. MGRMPL
96-well Microplate Magnetic Separation Rack
- Designed to be used with 96-well microplates, ELISA plates
- 96 magnets are positiont below microplate wells for efficient magnetic separation
- Can be used for high throughput separation of up to 96 samples
- Housing material: PMMA/Acrylic
- Magnet: N38 Neodymium (NdFeB)