Learn More
Molecular Devices Molecular Devices Picking Head, 96-Pin, E. coli, 96 and 384-Well
Catalog No. 5000014354
Encompass_Preferred
Molecular Devices X4006G is a 96-pin picking head engineered for automated colony picking of E. coli on 400-series colony picking systems. It mounts on compatible pickers and uses dedicated picking pins to transfer colonies reliably from agar plates to microplates.
  • Optimized for picking E. coli colonies.
  • 96-pin configuration for high-throughput colony transfer.
  • Compatible with standard 96-well and 384-well plates, including deep-well formats.
  • Uses picking pins with 0.55 mm tip diameter for precise colony sampling.
  • Designed for use with 400-series automated colony picking systems.
Catalog No. 50-000-14354 Supplier Molecular Devices Supplier No. X4006G
Add to Cart
Add to Cart
Request a Quote
Molecular Devices X4006G is a 96-pin picking head engineered for automated colony picking of E. coli on 400-series colony picking systems. It mounts on compatible pickers and uses dedicated picking pins to transfer colonies reliably from agar plates to microplates.
  • Optimized for picking E. coli colonies.
  • 96-pin configuration for high-throughput colony transfer.
  • Compatible with standard 96-well and 384-well plates, including deep-well formats.
  • Uses picking pins with 0.55 mm tip diameter for precise colony sampling.
  • Designed for use with 400-series automated colony picking systems.