Learn More
Molecular Devices Molecular Devices Microplate Reader, Multi-Mode, 8-Channel, 2 to 45 °C
Catalog No. 5000014711
Encompass_Preferred
The FlexStation 3 Multi-Mode Microplate Reader is a benchtop multimode instrument for kinetic and endpoint assays with integrated liquid handling and temperature control. It measures absorbance, fluorescence intensity, fluorescence polarization, luminescence, and time-resolved fluorescence using monochromators for flexible wavelength selection.
  • Integrated 8-channel pipettor for timed reagent additions.
  • Supports 6, 12, 24, 48, 96, and 384 well plate formats.
  • Detection modes include absorbance, fluorescence intensity, fluorescence polarization, luminescence, and time-resolved fluorescence.
  • Monochromators tunable in 1.0 nm increments for flexible wavelength selection.
  • Wavelength coverage suitable for absorbance and fluorescence assays.
  • Temperature control from 2°C above ambient to 45°C for incubated assays.
  • Suitable for kinetic assays, compound additions, and high-throughput workflows.
Catalog No. 50-000-14711 Supplier Molecular Devices Supplier No. FLEX38G
Add to Cart
Add to Cart
Request a Quote
The FlexStation 3 Multi-Mode Microplate Reader is a benchtop multimode instrument for kinetic and endpoint assays with integrated liquid handling and temperature control. It measures absorbance, fluorescence intensity, fluorescence polarization, luminescence, and time-resolved fluorescence using monochromators for flexible wavelength selection.
  • Integrated 8-channel pipettor for timed reagent additions.
  • Supports 6, 12, 24, 48, 96, and 384 well plate formats.
  • Detection modes include absorbance, fluorescence intensity, fluorescence polarization, luminescence, and time-resolved fluorescence.
  • Monochromators tunable in 1.0 nm increments for flexible wavelength selection.
  • Wavelength coverage suitable for absorbance and fluorescence assays.
  • Temperature control from 2°C above ambient to 45°C for incubated assays.
  • Suitable for kinetic assays, compound additions, and high-throughput workflows.