Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Integrated Pipettor, Dual Monochromators, 6-384 Well
Catalog No. 5000014732
Encompass_Preferred
The FlexStation 3 is an integrated benchtop microplate reader that combines multi-mode detection with programmable liquid handling to automate biochemical and cell-based assays and support kinetic measurements.
  • Automated 8- or 16-channel pipettor for reagent transfer.
  • Measures absorbance, fluorescence intensity, fluorescence polarization, luminescence, and time-resolved fluorescence.
  • Supports 6-384 well plate formats.
  • Dual monochromators with 1-nm wavelength selection.
  • Top and bottom reading capability with PMT and silicon photodiode detectors.
  • SoftMax Pro compatible acquisition and analysis software.
  • Compact benchtop footprint, approximately 48 x 58 x 41 cm.
Catalog No. 50-000-14732 Supplier Molecular Devices Supplier No. FLEX3
Add to Cart
Add to Cart
Request a Quote
The FlexStation 3 is an integrated benchtop microplate reader that combines multi-mode detection with programmable liquid handling to automate biochemical and cell-based assays and support kinetic measurements.
  • Automated 8- or 16-channel pipettor for reagent transfer.
  • Measures absorbance, fluorescence intensity, fluorescence polarization, luminescence, and time-resolved fluorescence.
  • Supports 6-384 well plate formats.
  • Dual monochromators with 1-nm wavelength selection.
  • Top and bottom reading capability with PMT and silicon photodiode detectors.
  • SoftMax Pro compatible acquisition and analysis software.
  • Compact benchtop footprint, approximately 48 x 58 x 41 cm.