Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Dual Monochromator, 6-384-Well
Catalog No. 5000014392
Encompass_Preferred
The SpectraMax M3 multi-mode microplate reader is a configurable, high-performance instrument for absorbance, fluorescence, and luminescence assays, designed for both research and regulated laboratory environments. It combines tunable optics with flexible plate handling and compliance-capable analysis software.
  • Dual monochromator optics, tunable in 1-nm increments
  • Detection modes include absorbance, fluorescence, luminescence, fluorescence polarization, and time-resolved assays
  • Supports 6- to 384-well microplates, with top and bottom reading options
  • High-sensitivity detectors with xenon flash lamp illumination
  • Software features secure data acquisition and 21 CFR Part 11-compatible controls
Catalog No. 50-000-14392 Supplier Molecular Devices Supplier No. M3
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M3 multi-mode microplate reader is a configurable, high-performance instrument for absorbance, fluorescence, and luminescence assays, designed for both research and regulated laboratory environments. It combines tunable optics with flexible plate handling and compliance-capable analysis software.
  • Dual monochromator optics, tunable in 1-nm increments
  • Detection modes include absorbance, fluorescence, luminescence, fluorescence polarization, and time-resolved assays
  • Supports 6- to 384-well microplates, with top and bottom reading options
  • High-sensitivity detectors with xenon flash lamp illumination
  • Software features secure data acquisition and 21 CFR Part 11-compatible controls