Learn More
Molecular Devices Molecular Devices Microplate Reader, Multi-Mode, Dual Monochromators, 6-384 Well
Catalog No. 5000014391
Encompass_Preferred
The SpectraMax M5 multi-mode microplate reader delivers flexible, sensitive measurement capabilities for a wide range of plate-based biochemical and cell-based assays. It combines scanning monochromators, automated detector optimization, and an integrated cuvette port to support absorbance, fluorescence, luminescence, and time-resolved assays across 6-384 well plates.
  • Multi-mode detection for absorbance, fluorescence, luminescence, TRF, and FP.
  • Dual scanning monochromators with 1 nm wavelength resolution.
  • AutoPMT optimization for extended dynamic range and per-well sensitivity.
  • Integrated cuvette port for single-cuvette measurements.
  • Temperature control from ambient +2°C to ~60°C with linear shaking.
  • Supports 6-384 well plates and automation integration.
Catalog No. 50-000-14391 Supplier Molecular Devices Supplier No. M5
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M5 multi-mode microplate reader delivers flexible, sensitive measurement capabilities for a wide range of plate-based biochemical and cell-based assays. It combines scanning monochromators, automated detector optimization, and an integrated cuvette port to support absorbance, fluorescence, luminescence, and time-resolved assays across 6-384 well plates.
  • Multi-mode detection for absorbance, fluorescence, luminescence, TRF, and FP.
  • Dual scanning monochromators with 1 nm wavelength resolution.
  • AutoPMT optimization for extended dynamic range and per-well sensitivity.
  • Integrated cuvette port for single-cuvette measurements.
  • Temperature control from ambient +2°C to ~60°C with linear shaking.
  • Supports 6-384 well plates and automation integration.