Learn More
Molecular Devices Molecular Devices Microplate Reader, Multi-Mode, 200-1000 nm, 90-240 V
Catalog No. 5000014715
Encompass_Preferred
High-performance multi-mode microplate reader for absorbance, fluorescence, and luminescence assays. Supports plate and cuvette measurements, automated workflows, and controlled-temperature experiments.
  • Multi-mode detection including absorbance, fluorescence, luminescence, TRF, FP, and HTRF.
  • Wavelength ranges: absorbance 200-1000 nm, fluorescence and luminescence 250-850 nm.
  • Xenon flashlamp illumination with photomultiplier tube and silicon photodiode detection.
  • Temperature control from ambient +2°C to 60°C.
  • Automation ready with cuvette port, plate shaking, and robotics compatibility.
  • Power input 90-240 V AC.
Catalog No. 50-000-14715 Supplier Molecular Devices Supplier No. MINIMAX300
Add to Cart
Add to Cart
Request a Quote
High-performance multi-mode microplate reader for absorbance, fluorescence, and luminescence assays. Supports plate and cuvette measurements, automated workflows, and controlled-temperature experiments.
  • Multi-mode detection including absorbance, fluorescence, luminescence, TRF, FP, and HTRF.
  • Wavelength ranges: absorbance 200-1000 nm, fluorescence and luminescence 250-850 nm.
  • Xenon flashlamp illumination with photomultiplier tube and silicon photodiode detection.
  • Temperature control from ambient +2°C to 60°C.
  • Automation ready with cuvette port, plate shaking, and robotics compatibility.
  • Power input 90-240 V AC.