Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Aluminum Support, tube/plate rack, 96-well

Catalog No. FERPRK1963 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 rack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERPRK1963 1 rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERPRK1963 Supplier Thermo Scientific™ Supplier No. PRK1963
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ aluminum support rack can be used for sample preparation, heat-sealing and automatic dispensing.

Perform sample preparation, heat-sealing and automatic dispensing using Thermo Scientific™ aluminum support rack.

Available in 96- and 384-well format.

  • For heat-sealing and automated dispensing
  • Industry-standard footprint fits in robotic workstations
  • Can be heated or cooled to assist with sample preparation

Includes:

Recommended for:
Sample handling, sample preparation, heat-sealing, automated dispensing

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Holds Microplates (-well)
Material Aluminum
Shape Rectangular
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Quantity 1 rack
Product Type Microplate Rack

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.