Learn More
JDM LAB SOLUTIONS LLC MICROPLATE SHAKER
Catalog No. NC2307316
Encompass
Microplate Shaker OS-07UAFeatures Ergonomic design with speed control and timer Automatically recovers from power outages Wide range of accessories available Digital control and LED/LCD display Designed and built for continuous use Gentle speed up and slow down Safe for use inside standard incubators No vibration or heat output Built for heavy workloadsSpeed: 150 - 1,000 RPMCapacity: 4 microplates
Catalog No. NC2307316 Supplier JDM LAB SOLUTIONS LLC Supplier No. OS07UA
Microplate Shaker OS-07UAFeatures Ergonomic design with speed control and timer Automatically recovers from power outages Wide range of accessories available Digital control and LED/LCD display Designed and built for continuous use Gentle speed up and slow down Safe for use inside standard incubators No vibration or heat output Built for heavy workloadsSpeed: 150 - 1,000 RPMCapacity: 4 microplates