Learn More
Fine Science Tools Micro Points, Angled 90° Long, 15.5 cm
SDP
Catalog No. NC9773566
Encompass
Product: Micro Point - Angled 90 deg. Long/15.5cm/0.02mm Tip Diameter/ Tip Shape: Angled 90 deg. Long/ Tip Diameter: 0.02mm/ Alloy: Stainless Steel15.5cm/ Feature: Each
Catalog No. NC9773566 Supplier Fine Science Tools Supplier No. 1006515
Product: Micro Point - Angled 90 deg. Long/15.5cm/0.02mm Tip Diameter/ Tip Shape: Angled 90 deg. Long/ Tip Diameter: 0.02mm/ Alloy: Stainless Steel15.5cm/ Feature: Each