Learn More
Research Products International Corp Micro-Point Scissors, 3/4 Inch Blade, Sharp-Sharp-Straight, 5 x 1 x 1/2 Inches
SDP
Catalog No. 50212531
Encompass
MicroPoint Scissors are made from high quality (memory) stainless steel and designed to cut by an adjustable shearing action. The increased sheer capacity of the MicroPoint allows a "micro-point" of shearing contact throughout the entire shear. This allows a sharper cut and guarantees consistent shearing contact, from the rear of the scissors to the point. Ideal for producing a clean cut at actual pressure point. Supplied with protective shield for added safety and lifetime guarantee against corrosion and defects.
Three year warranty.
Catalog No. 50-212-531 Supplier Research Products International Corp Supplier No. 247200
MicroPoint Scissors are made from high quality (memory) stainless steel and designed to cut by an adjustable shearing action. The increased sheer capacity of the MicroPoint allows a "micro-point" of shearing contact throughout the entire shear. This allows a sharper cut and guarantees consistent shearing contact, from the rear of the scissors to the point. Ideal for producing a clean cut at actual pressure point. Supplied with protective shield for added safety and lifetime guarantee against corrosion and defects.
Three year warranty.