Learn More
Research Products International Corp MicroPoint Scissors, 3/8"", Micro-Tenotomy
SDP
Catalog No. 501978091
Encompass
MicroPoint scissors can be used for suture or tissue, and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors do not cut by cutting. They cut by shearing.
Catalog No. 50-197-8091 Supplier Research Products International Corp Supplier No. 247205
MicroPoint scissors can be used for suture or tissue, and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors do not cut by cutting. They cut by shearing.