Learn More
Electron Microscopy Sciences MicroPol™ MC3 Polisher For TEM & Metallography
SDP
Catalog No. 50949933
Encompass
The MicroPol Polisher is a compact, versatile, electrically controlled, precise mechanical specimen grinder/polisher designed for planar grinding, thinning and polishing of specimen in the fields of nanotechnology, semiconductors, and materials science. The parameters of polishing (speed, load, time, etc.) can be matched according to the properties of the specimen. High quality samples can be prepared with minimal effort by the operator.
Features rugged, corrosion proof construction, variable speed control, quick change bayonet mount bowls, timer for automatic operation, and extremely light load for fragile TEM Samples.
While the Micropol™ MC3 is well suited for metallographic polishing, it has been specially designed for precision TEM polishing applications. Two types of specimen holders are provided with each unit that makes it ideal for the preparation of both planar and dimpled samples, respectively.
Catalog No. 50-949-933 Supplier Electron Microscopy Sciences Supplier No. 50174MC2
Add to Cart
Add to Cart
Request a Quote
The MicroPol Polisher is a compact, versatile, electrically controlled, precise mechanical specimen grinder/polisher designed for planar grinding, thinning and polishing of specimen in the fields of nanotechnology, semiconductors, and materials science. The parameters of polishing (speed, load, time, etc.) can be matched according to the properties of the specimen. High quality samples can be prepared with minimal effort by the operator.
Features rugged, corrosion proof construction, variable speed control, quick change bayonet mount bowls, timer for automatic operation, and extremely light load for fragile TEM Samples.
While the Micropol™ MC3 is well suited for metallographic polishing, it has been specially designed for precision TEM polishing applications. Two types of specimen holders are provided with each unit that makes it ideal for the preparation of both planar and dimpled samples, respectively.