Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire™ MicroPolx™ 2000LT Sealed Edge Cleanroom Wiper

Catalog No. 19161312 Shop All Berkshire Corporation Products
Change view
Click to view available options
Dimensions (L x W):
22.86 x 22.86 cm (9 x 9 in.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-161-312 22.86 x 22.86 cm (9 x 9 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-161-312 Supplier Berkshire™ Supplier No. MP2000LT0910
This item is not returnable. View return policy

Designed for use in ISO Class 4 (Class 10) and higher cleanroom environments. Berkshire MicroPolx 2000LT Sealed Edge Cleanroom Wipers are constructed of 80% polyester/20% nylon microfiber yarn that increases surface area and provides superior wiping efficiency.

  • Ultrafine, wedge-shaped polyester and nylon filaments, tight knit construction, and a soft, smooth surface provide superior particle and bacterial removal
  • 16 times the number of filaments and 6 times the surface area of standard polyester wipers
  • Four sided thermally sealed edges to minimize fiber and particle generation
  • Contains low Titanium dioxide (TiO2) to minimize cleanroom contamination
  • Cleans efficiently with less solvent usage than traditional wipers
  • Removes oily soils with little or no solvent required
  • Soft hand and non-abrasive fabric surface is excellent for sensitive or scratch prone surfaces
  • Chemically compatible with IPA and other commonly used solvents

Specifications

Product Type Cleanroom Wiper
Material Polyester, Nylon
Color White
Edges Sealed
Dimensions (L x W) 22.86 x 22.86 cm (9 x 9 in.)
For Use With (Application) Cleaning sensitive surfaces, precise optical machinery and tools, delicate process in critical cleanrooms used in display and semiconductor industries and microelectronics manufacturing
Packaging Quantity 100 Sheets/Pk., 10 Pk./Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.