Learn More
Valutek™ Microporous Coverall with Full Zipper, Elastic Cuff & Ankle
Description
- All coveralls have a flap over zipper closure, elastic wrist, and ankle cuffs design
- Available with either a single ply shirt collar or with an elastic hood and attached anti-skid PVC sole boot covers
- The coveralls are highly breathable and provide the operators with comfortable full body coverage protection
- All Valutek™ coveralls are tested and manufactured in ISO compliant facilities under Valutek™ inspection and stringent process control to ensure Valutek™ quality standard and product specification compliance
- This product is recommended for either stringent industrial or demanding cleanroom applications
- This coverall is ideal for use in a cleanroom Class 1-10 (ISO 3-4) critical environment as well as other critical areas that require high contamination control and cleanliness
Specifications
Specifications
| Size | Small |
| Disposable | Single-use |
| Cuff Style | Elastic |
| Closure Type | Zipper |
| Sterility | Non-sterile |
| Resistance | Non-toxic Particulates |