Learn More
Valutek™ Microporous Coverall with Full Zipper, Elastic Cuff & Ankle
Catalog No. 191502104
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-2104 4X-Large
19-150-2098 Small
19-150-2099 Medium
19-150-2100 Large
19-150-2101 X-Large
19-150-2102 2X-Large
19-150-2103 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-150-2104 Supplier Valutek™ Supplier No. VTMCVRL4X
Add to Cart
Add to Cart
This item is not returnable. View return policy
Disposable microporous coveralls are manufactured with 100% non-linting microporous film bonded with a non-woven fabric to provide superior barrier protection against non-toxic particulates, liquids, spray and dust
  • All coveralls have a flap over zipper closure, elastic wrist, and ankle cuffs design
  • Available with either a single ply shirt collar or with an elastic hood and attached anti-skid PVC sole boot covers
  • The coveralls are highly breathable and provide the operators with comfortable full body coverage protection
  • All Valutek™ coveralls are tested and manufactured in ISO compliant facilities under Valutek™ inspection and stringent process control to ensure Valutek™ quality standard and product specification compliance
  • This product is recommended for either stringent industrial or demanding cleanroom applications
  • This coverall is ideal for use in a cleanroom Class 1-10 (ISO 3-4) critical environment as well as other critical areas that require high contamination control and cleanliness
Specifications
Size 4X-Large
Disposable Single-use
Cuff Style Elastic
Closure Type Zipper
Sterility Non-sterile
Resistance Non-toxic Particulates