Learn More
MicroGuard MP™ Microporous Coveralls, Elastic Wrists and Open Ankles
Catalog No. 19472757 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-757 X-Large
19-472-756 Small
19-472-755 Medium
19-472-754 Large
19-472-750 2X-Large
19-472-751 3X-Large
19-472-752 4X-Large
19-472-753 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-472-757 Supplier MicroGuard MP™ Supplier No. 8012XL
Add to Cart
Add to Cart
Microporous coveralls deliver protection and comfort through a strong and durable fabric.
  • Comfortable state-of-the-art lamination technology allows for maximum breathability.
  • Soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so heat and moisture are not trapped near the body.
  • Elastic wrists and open ankles.
  • Coveralls offer base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection.
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas.
  • Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric.
  • Elastic back stretches with user for more comfort and better tear resistance.
  • Elastic wrists and ankles offer a secure fit to help prevent skin exposure.
  • Environmentally friendly, can be recycled after use.
  • Attached hood provides additional protection of the neck and shoulders.
Specifications
Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Open
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class General
Resistance Abrasion, Caustics, Particle, Liquid, Water
Back Style Elastic
Model 8012
No. of Pockets None