Learn More
MSC MICROPOROUS FILM DISP SLV
Catalog No. 5000792003
Encompass
These PRO-SAFE disposable sleeves are designed for protective apparel applications, offering a barrier against particulates and liquids. Made from microporous material, they provide comfort and breathability while maintaining protection.
  • Disposable sleeve with elastic opening at both ends
  • Constructed from microporous material for protection and breathability
  • Provides coverage for arms in various work environments
  • Size L, 18 inch long sleeve
Catalog No. 50-007-92003 Supplier MSC Supplier No. 73601783
These PRO-SAFE disposable sleeves are designed for protective apparel applications, offering a barrier against particulates and liquids. Made from microporous material, they provide comfort and breathability while maintaining protection.
  • Disposable sleeve with elastic opening at both ends
  • Constructed from microporous material for protection and breathability
  • Provides coverage for arms in various work environments
  • Size L, 18 inch long sleeve