Learn More
Valutek™ Microporous Hood
Catalog No. 191502085
Change view
Click to view available options
Quantity:
100/Pk.
Catalog No. Quantity
19-150-2085 100/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-2085 Supplier Valutek™ Supplier No. VTMHOODU
Add to Cart
Add to Cart
This item is not returnable. View return policy
Manufactured with 100% non-linting microporous film bonded with a non-woven fabric. Provides protection against non-toxic particulates, liquids, spray, and dust.
  • Hoods are tested and manufactured in ISO compliant facilities under inspection and stringent process control to ensure quality standard and product specification compliance
  • Recommended for either stringent industrial or demanding cleanroom applications
  • Ideal for use in a cleanroom Class 1-10 (ISO 3-4) critical environment that requires high contamination control and cleanliness
Specifications
Product Type Open Face
Color White
Closure Type None
Disposable Single-use
Model VTMHOOD-U
Sterility Non-sterile
Quantity 100/Pk.
For Use With (Application) Recommended for either stringent industrial or demanding cleanroom applications. This hood is ideal for use in a cleanroom Class 1-10 (ISO 3-4) critical environment that requires high contamination control and cleanliness.
Cleanroom Class Class 1 (ISO 3)