Learn More
International Enviroguard™ MicroGuard MP™ Microporous Lab Coats, No Pockets
Catalog No. 19472454 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-454 Medium
19-472-455 Large
19-472-456 X-Large
19-472-457 2X-Large
19-472-458 3X-Large
19-472-459 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-472-454 Supplier International Enviroguard™ Supplier No. 8046M
Add to Cart
Add to Cart
Microporous lab coats deliver protection and comfort through a strong and durable fabric designed for worker comfort.
  • Microporous, disposable lab coats offer the balance of safety and comfort
  • Fabric keeps dust and liquids out while allowing air to flow freely to keep worker cool
  • Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
  • Produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
  • Breathable fabric lets moisture vapor escape
  • No pockets
  • Folded collar
  • Environmentally friendly- can be recycled after use
  • Elastic wrists offer a snug, secure fit to keep sleeves in place to prevent skin exposure
Specifications
Product Type Controlled Environments Lab Coat
Size Medium
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Collar Type Folded
Seam Type Serged