Learn More
Lakeland Industries CLEANMAX CLEAN STERILE Microporous Cleanroom coverall with Hood. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. Individually packaged. 25 per case with overlay bag. Size 5X
Catalog No. NC2958046
Encompass_Preferred
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, microporous fabric. Built in hood, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. 25/case.
Catalog No. NC2958046 Supplier Lakeland Industries Supplier No. CTL428CS5X
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, microporous fabric. Built in hood, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. 25/case.