Learn More
USA Scientific Inc Microporous pierceable film
Catalog No. 502744839
Encompass
Microporous pierceable rayon film, -20C to +80C, non-str, 100/box
Catalog No. 50-274-4839 Supplier USA Scientific Inc Supplier No. 29201000
Microporous pierceable rayon film, -20C to +80C, non-str, 100/box