Learn More
World Precision Instrument Microprobe Thermometers -BAT-12R-220
SDP
Catalog No. 502538918
Encompass
A Microprobe Thermometer is the instrument of choice for biological and laboratory temperature measurements. These thermometers are very versatile, providing fast response, high accuracy and stability with digital display and analog signal for connection to a computer or recorder. With the wide selection of probes, the instruments can be used in almost any application.
Catalog No. 50-253-8918 Supplier World Precision Instrument Supplier No. BAT12R220
A Microprobe Thermometer is the instrument of choice for biological and laboratory temperature measurements. These thermometers are very versatile, providing fast response, high accuracy and stability with digital display and analog signal for connection to a computer or recorder. With the wide selection of probes, the instruments can be used in almost any application.