Learn More
AVANTOR MASTERFLEX (VWR LLC) A-Mount Cavity Style Pump Head; SS/PEEK/PTFE; 0.64 mL/rev
Catalog No. NC1553090
Encompass
Micropump L18428 A-Mount Cavity Style Pump Head; SS, PEEK, PTFE; 0.64 mL/rev, Smooth pulseless flow provides precise, leak-free delivery of fluidsCavity-style pump heads are designed to handle higher inlet pressures.
Catalog No. NC1553090 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7301108
Micropump L18428 A-Mount Cavity Style Pump Head; SS, PEEK, PTFE; 0.64 mL/rev, Smooth pulseless flow provides precise, leak-free delivery of fluidsCavity-style pump heads are designed to handle higher inlet pressures.