Learn More
AVANTOR MASTERFLEX (VWR LLC) Micropump L20013 A-Mount Suction Shoe Pump Head; SS/PPS/PTFE; 0.092 mL/rev
Catalog No. NC1653785
Encompass
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Catalog No. NC1653785 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0700214
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.