Learn More
AVANTOR MASTERFLEX (VWR LLC) Micropump L20561 A-Mount Suction Shoe Pump Head; Bypass/SS/PEEK/Viton; 0.261 mL/rev
Catalog No. NC1659992
Encompass
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Catalog No. NC1659992 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7300400
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.