Learn More
Electron Microscopy Sciences Aluminium Micro-Punch, Size: 1.0 mm., 1/EA
SDP
Supplier:  Electron Microscopy Sciences 6903310
Encompass_Preferred
The EMS Aluminum Punch Kit is designed to serve research scientists with a dissection tool for small, pre-defined regions of the tissue or the specimen. Barrel is machined from high grade 6061 light weight, seamless, aluminium tubing and anodized to form a protective, chemically resistant surface. Easy handling. All punches are 130 mm in length, with a shaft of 3.2 mm and a tip length of 16.8 mm. Deluxe plastic case included. Size: 1.0 mm.
Catalog No. 50-209-0940